Merck
HPLC Analysis of Fat Soluble Vitamins on Ascentis® Express RP-Amide, Methanol Mobile Phase
HPLC Analysis of Fat Soluble Vitamins on Ascentis® Express RP-Amide, Methanol Mobile Phase application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Ascentis® Express RP-Amide, 2.7 μm HPLC Column
2.7 μm particle size, L × I.D. 15 cm × 4.6 mm
standard
Product No.
Description
Pricing
Cholecalciferol (D3)
analytical standard
Ergocalciferol (D2)
analytical standard
Phylloquinone (K1)
analytical standard
(±)-α-Tocopherol
analytical standard
DL-alpha-Tocopherol acetate
analytical standard
CONDITIONS
column
Ascentis Express RP-Amide, 15 cm x 4.6 mm I.D., 2.7 μm particles (53931-U)
mobile phase
methanol
flow rate
1.0 mL/min
pressure
1653 psi (114 bar)
column temp.
30 °C
detector
UV, 280 nm
injection
5 μL
sample
0.4 mg/mL tocopherol acetate, alpha-tocopherol and K1, 0.04 mg/mL D2 and D3, all in ethanol
Description
Analysis Note
Analysis was conducted using standard solution, detection via UV.
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany