HPLC Analysis of Asparaginase on Zenix® SEC-150
![HPLC Analysis of Asparaginase on Zenix® SEC-150 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/748/143/6acde95e-8903-424f-9861-c17975512219/800/6acde95e-8903-424f-9861-c17975512219.jpg)
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-150 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steelCONDITIONS
column
Zenix SEC-150, 30 cm x 7.8 mm I.D., 3 μm particles (Z777018)
mobile phase
150 mM sodium phosphate, pH 7.0
flow rate
0.6 mL/min
pressure
1015 psi (70 bar)
column temp.
ambient
detector
UV, 280 nm
inj. temp.
5 μL
sample
asparaginase (2.0 mg/mL in water), RT 10.895 minute, MW 5000-6000, pI 4.85
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies