US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis
![US EPA Method OLM04.2 VOA: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap, Fast GC Analysis suitable for GC](/deepweb/assets/sigmaaldrich/product/images/141/514/fed88219-139c-46dd-a51c-decf5ddfbf5b/800/fed88219-139c-46dd-a51c-decf5ddfbf5b.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
sample/matrix
each analyte at 50 ppb in 5 mL water
purge trap
VOCARB 3000 “K”
purge
40 mL/min at 25 °C for 11 min
dry purge
2 min
desorption preheat
205 °C
desorption process
124 mL/min. at 210 °C for 2 min.
bake
260 °C for 10 min
transfer line/valve temp.
110 °C
column
SPB-624, 20 m × 0.18 mm I.D., 1.0 μm (28662-U)
oven
40 °C (1 min), 11 °C/min to 125 °C, 35 °C/min to 230 °C (2 min)
inj. temp.
150 °C
MSD interface
200 °C
scan range
m/z = 35-400
carrier gas
helium, 1.2 mL/min
injection
100:1 split
liner
0.75 mm I.D. SPME
Description
Legal Information
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany