Merck
GC Analysis of 1-Aminoindan Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX G-DA
GC Analysis of 1-Aminoindan Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX™ G-DA suitable for GC
Materials
standard
Product No.
Description
Pricing
1-Aminoindan
98%
(R)-(−)-1-Aminoindane
97%
(S)-(+)-1-Aminoindan
97%
(R)-(−)-1-Aminoindane
ChiPros®, produced by BASF, 99%
CONDITIONS
column
Astec CHIRALDEX G-DA, 30 m x 0.25 mm I.D., 0.12 μm (72033AST)
oven
150 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 24 psi
sample
peaks 1 & 2: 1-aminoindan enantiomers (N-TFA derivatives)
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC