US EPA Method 8270 (Appendix IX): GC Analysis of Semivolatiles on Equity-5 (30 m x 0.25 mm I.D., 0.50 μm)
CONDITIONS
column
Equity-5, 30 m × 0.25 mm I.D., 0.5 μm (28092-U)
oven
40 °C (2 min), 50 °C/min to 100 °C, 10 °C/min to 200 °C, 30 °C/min to 325 °C (7.5 min)
inj. temp.
280 °C, splitless (0.61 min)
detector
MSD, Scan range 45-450 amu, 325°C
carrier gas
helium, pressure programmed, 20 psi (0.0 min), ramp to 80 psi (0.0 min), ramp to 16.5 psi (3 min), ramp to 25 psi (hold for remainder of run)
injection
1 μL, splitless (0.61 min)
liner
4 mm I.D., single taper
sample
50 ng on-column of a 62 Supplemental Appendix IX list consisting of semivolatile standards, 6 internal standards, and 8 surrogates
Description
Legal Information
null