GC Analysis of Conjugated Linoleic Acid (CLA) FAME Isomers on SLB®-IL111
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
SLB-IL111, 200 m x 0.25 mm I.D., 0.20 μm (29689-U)
oven
150 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
hydrogen
injection
1 μL 1μL, split 50:1
liner
4 mm I.D., split type, cup design
sample
4-component CLA FAME isomer mix, 0.5 mg/mL in methylene chloride
Description
Analysis Note
Four key conjugated linoleic acid (CLA) FAME isomers are resolved. The SLB-IL111 provides a different elution pattern compared to SP-2560 (a cyanopropyl siloxane column).
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany