Merck
GC Analysis of Class 1 Residual Solvents on Equity-1 after SPME using 100 μm PDMS Fiber
GC Analysis of Class 1 Residual Solvents on Equity-1 after SPME using 100 μm PDMS Fiber suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 10 m × 0.20 mm, df 1.20 μm
SPME fiber
Product No.
Description
Pricing
SPME Fiber Assembly
100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, plain red hub
CONDITIONS
sample/matrix
2 mL water + sodium chloride at 25% containing each analyte at 1 ppm, in a 4 mL vial
SPME fiber
100 μm PDMS, 24 ga
extraction
heated headspace, 50 °C for 5 min
desorption process
250 °C for 3 min
column
Equity-1, 10 m x 0.20 mm I.D., 1.2 μm (28043-U)
oven
40 °C (0.75 min), 20 °C/min to 200 °C (10 min)
inj. temp.
250 °C
detector
FID
carrier gas
helium, 35 cm/sec @ 40 °C
injection
5:1 split
liner
0.75 mm I.D. SPME type
Description
Legal Information
null