Merck
GC Analysis of Allergens in Perfume on SLB®-5ms (5 m x 0.05 mm I.D., 0.05 μm), Fast GC Analysis
GC Analysis of Allergens in Perfume on SLB®-5ms (5 m x 0.05 mm I.D., 0.05 μm), Fast GC Analysis suitable for GC
CONDITIONS
column
SLB-5ms, 5 m x 0.05 mm I.D., 0.05 μm
oven
40 °C, 70 °C/min to 300 °C
detector
FID, 305 °C
inj. temp.
290 °C
carrier gas
hydrogen, 80 cm/sec, 554.4 kPa
injection
0.2 μL, 250:1 split
Description
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany