Merck
GC Analysis of a 37-Component FAME Mix on SP®-2560, Slower Oven Ramp
GC Analysis of a 37-Component FAME Mix on SP®-2560, Slower Oven Ramp suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SP®-2560 Capillary GC Column
L × I.D. 100 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SP-2560, 100 m × 0.25 mm I.D., 0.20 μm (24056)
oven
140 °C (5 min) to 240 °C (5 min) at 4 °C/min
detector
FID, 260 °C
carrier gas
helium, 20 cm/sec
sample volume
1 μL split 100:1, 260 °C
Description
Legal Information
SP is a registered trademark of Sigma-Aldrich Co. LLC