Merck
  • The ERK/MAPK pathway regulates the activity of the human tissue factor pathway inhibitor-2 promoter.
The ERK/MAPK pathway regulates the activity of the human tissue factor pathway inhibitor-2 promoter.
The Journal of biological chemistry (2002-11-26)
Christina Kast, Minglun Wang, Malcolm Whiteway
ABSTRACT
Human tissue factor pathway inhibitor-2 (hTFPI-2) is a 32-kDa serine protease inhibitor that is associated with the extracellular matrix. hTFPI-2 inhibits several extracellular matrix-degrading serine proteases and may play a role in tumor invasion and metastasis. To study the signal transduction pathway that leads to the activation of the hTFPI-2, we cloned the potential promoter region of this gene adjacent to a heterologous luciferase reporter gene. Phorbol 12-myristate 13-acetate (PMA) induced the luciferase reporter gene in HEK293 cells and other epithelial cell lines, such as the human lung carcinoma A549 cells, the breast carcinoma MCF7 cells, and the cervical HeLa cells. This PMA induction was blocked with the MEK inhibitor UO126, suggesting that the PMA-induced activation of the hTFPI-2 promoter is mediated through MEK. Furthermore, epidermal growth factor induced the luciferase reporter gene in HeLa cells. Cotransfection of the luciferase construct with constitutively active components of the Ras/Raf/MEK/ERK pathway in EcR-293 cells lead to a 7- to 92-fold induction of the luciferase reporter gene, indicating that regulation of hTFPI-2 is mediated through this pathway. A series of luciferase reporter gene constructs with progressive deletions of the 5'-flanking region suggested that the minimal basal promoter activity is located between nucleotide positions -89 and -384, whereas the minimal inducible promoter activity is between -89 and -222. We have used the computer program TFSEARCH and mutagenesis to analyze potential transcription factor binding sites. We identified an AP-1 binding site at nucleotide position -156 (inducible activity) and a Sp1 site at position -134 (basal activity) as potential cis-acting elements in the promoter region of the hTFPI-2.