Merck
  • Association of HLA-A, B, DRB1 alleles and haplotypes with HIV-1 infection in Chongqing, China.
Association of HLA-A, B, DRB1 alleles and haplotypes with HIV-1 infection in Chongqing, China.
BMC infectious diseases (2009-12-17)
Xia Huang, Hua Ling, Wei Mao, Xianbin Ding, Quanhua Zhou, Mei Han, Fang Wang, Lei Cheng, Hongyan Xiong
ABSTRACT
The human immunodeficiency virus type 1(HIV-1) epidemic in Chongqing, China, is increasing rapidly with the dominant subtype of CRF07_BC over the past 3 years. Since human leukocyte antigen (HLA) polymorphisms have shown strong association with susceptibility/resistance to HIV-1 infection from individuals with different ethnic backgrounds, a recent investigation on frequencies of HLA class I and class II alleles in a Chinese cohort also indicated that similar correlation existed in HIV infected individuals from several provinces in China, however, such information is unavailable in Chongqing, southwest China. In this population-based study, we performed polymerase chain reaction analysis with sequence-specific oligonucleotide probes (PCR-SSOP) for intermediate-low-resolution HLA typing in a cohort of 549 HIV-1 infected individuals, another 2475 healthy subjects from the Han nationality in Chongqing, China, were selected as population control. We compared frequencies of HLA-A, B, DRB1 alleles, haplotypes and genotypes between the two groups, and analyzed their association with HIV-1 susceptibility or resistance. The genetic profile of HLA (A, B, DRB1) alleles of HIV-1 infected individuals from Chongqing Han of China was obtained. Several alleles of HLA-B such as B*46 (P = 0.001, OR = 1.38, 95%CI = 1.13-1.68), B*1501G(B62) (P = 0.013, OR = 1.42, 95%CI = 1.08-1.88), B*67 (P = 0.022, OR = 2.76, 95%CI = 1.16-6.57), B*37 (P = 0.014, OR = 1.93, 95%CI = 1.14-3.28) and B*52 (P = 0.038, OR = 1.64, 95%CI = 1.03-2.61) were observed to have association with susceptibility to HIV-1 infection in this population. In addition, the haplotype analysis revealed that A*11-B*46, A*24-B*54 and A*01-B*37 for 2-locus, and A*11-B*46-DRB1*09, A*02-B*46-DRB1*08, A*11-B*4001G-DRB1*15, A*02-B*4001G-DRB1*04, A*11-B*46-DRB1*08 and A*02-B*4001G-DRB1*12 for 3-locus had significantly overrepresented in HIV-1 infected individuals, whereas A*11-B*1502G, A*11-B*1502G-DRB1*12 and A*33-B*58-DRB1*13 were underrepresented. However, the low-resolution homozygosity of HLA-A, B, DRB1 loci and HLA-Bw4/Bw6 genotypes did not differ significantly between the two groups. These results may contribute to the database of HLA profiles in HIV-1 infected Chinese population, consequently, the association of certain HLA alleles with susceptibility or resistance to HIV-1 infection would provide with clues in choosing proper preventive strategies against HIV-1 infection and developing effective HIV-1 vaccines in Chinese population, especially for those in southwest China.