Merck
  • Liquid chromatographic determination of six antiepileptic drugs and two metabolites in microsamples of human plasma.
Liquid chromatographic determination of six antiepileptic drugs and two metabolites in microsamples of human plasma.
Therapeutic drug monitoring (1999-10-16)
K M Matar, P J Nicholls, A Tekle, S A Bawazir, M I Al-Hassan
ABSTRACT
A simple, rapid, sensitive, and reproducible high-performance liquid chromatographic (HPLC) method for simultaneous determination of the antiepileptic drugs (ethosuximide, primidone, lamotrigine, phenobarbital, phenytoin, and carbamazepine) and two metabolites (carbamazepine-diol and carbamazepine-epoxide) in human plasma is described. The procedure involves extraction of the drugs from human plasma (100 microL) with ether using 9-hydroxymethyl-10-carbamyl acridan as an internal standard. The extract was evaporated and reconstituted with mobile phase and then injected onto the chromatograph. The drugs and the internal standard were eluted from a Supelcosil LC-18 stainless steel column at ambient temperature with a mobile phase consisting of a 0.01M phosphate buffer/methanol/acetonitrile (65/18/17, v/v/v) adjusted to a pH of 7.5 with phosphoric acid and a flow rate of 1 mL/min. The effluent was monitored at 220 nm. Quantitation was achieved by using peak area ratio of each drug to the internal standard. The intraassay and interassay coefficients of variation (CV) ranged from 2.43% to 6.25% and from 3.02% to 5.85%, respectively. The absolute (extraction) and relative (analytical) recoveries for the drugs ranged from 70.7% to 104.4% and from 88.3% to 106.1%, respectively. Stability tests showed that the drugs were stable in plasma for at least 4 weeks when stored at -20 degrees C. The method was applied clinically for monitoring the AEDs in epileptic patients.
Product Description
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 3 mm
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Supelco
SUPELCOSIL LC-18 Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 2.1 mm
Supelco
SUPELCOSIL LC-18-S HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm