Merck
  • Headspace solid-phase microextraction-gas chromatography--mass spectrometry analysis of the volatile compounds of Evodia species fruits.
Headspace solid-phase microextraction-gas chromatography--mass spectrometry analysis of the volatile compounds of Evodia species fruits.
Journal of chromatography. A (2005-09-01)
Federica Pellati, Stefania Benvenuti, Fumihiko Yoshizaki, Davide Bertelli, Maria Cecilia Rossi
ABSTRACT
In this study the investigation of the aroma compounds of dried fruits of Evodia rutaecarpa (Juss.) Benth. and E. rutaecarpa (Juss.) Benth. var. officinalis (Dode) Huang (i.e. E. officinalis Dode) (Rutaceae family) was carried out to identify the odorous target components responsible for the characteristic aroma of these valuable natural products. To avoid the traditional and more time-consuming hydrodistillation, the analyses were carried out by means of headspace solid-phase microextraction (HS-SPME) coupled to gas chromatography-mass spectrometry (GC-MS). The SPME headspace volatiles were collected using a divinylbenzene-carboxen-polydimethylsiloxane (DVB-CAR-PDMS) fiber. The extraction conditions were optimized using a response surface experimental design to analyze the effect of three factors: extraction temperature, equilibrium time and extraction time. The best response was obtained when the extraction temperature was around 80 degrees C, equilibrium time near 25 min and extraction time close to 18 min. Analyses were performed by GC-MS with a 5% diphenyl-95% dimethyl polysiloxane (30 m x 0.25 mm I.D., film thickness 0.25 microm) capillary column using He as the carrier gas and a programmed temperature run. The main components of the HS-SPME samples of E. rutaecarpa (concentration >3.0%) were limonene (33.79%), beta-elemene (10.78%), linalool (8.15%), myrcene (5.83%), valencene (4.73%), beta-caryophyllene (4.62%), linalyl acetate (4.13%) and alpha-terpineol (3.99%). As for E. officinalis, the major compounds were myrcene (32.79%), limonene (18.36%), beta-caryophyllene (9.92%), trans-beta-ocimene (6.04%), linalool (5.88%), beta-elemene (7.85%) and valencene (4.62%).
Product Description
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
Tenax® TA / Carboxen® 1018, glass TD tube, preconditioned, O.D. × L 1/4 in. × 3 1/2 in., Sealed with (Swagelok® End-Fittings), pkg of 10 ea
Supelco
Carboxen® 569, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
ORBO 78 HBr on Carboxen® 564 specially cleaned (20/45), 400/200 mg, W,W,W separators, O.D. × L 6 mm × 110 mm, pkg of 25 ea
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 10 g
Supelco
Carboxen®-1010 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 60-80 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1003, 40-60 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 1000, 40-60 mesh, bottle of 50 g
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.32 mm, average thickness 15 μm
Supelco
Carboxen®-1006 PLOT Capillary GC Column, L × I.D. 30 m × 0.53 mm, average thickness 30 μm
Supelco
Carboxen® Adsorbent, matrix Carboxen® 563, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 564, 20-45 mesh, pack of 144 × 290 mg
Supelco
Carboxen® Adsorbent, matrix Carboxen® 572, 20-45 mesh, bottle of 10 g
Supelco
Carboxen® Adsorbent, matrix Carboxen® 569, 20-45 mesh, bottle of 500 g
Supelco
SPME Fiber Assembly, matrix active group 50/30µm DVB/CAR/PDMS, StableFlex fiber, manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 23 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, for use with manual holder, needle size 24 ga
Supelco
SPME Fiber Assembly, for use with autosampler, matrix active group 50/30µm DVB/CAR/PDMS, needle size 23 ga, metal alloy fiber, fiber L 2 cm
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 24 ga, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, for use with autosampler, needle size 23 ga, metal alloy fiber
Supelco
SPME Fiber Assembly, matrix active group 50/30µm DVB/CAR/PDMS, needle size 23 ga, metal alloy fiber, for use with autosampler, fiber L 1 cm, pkg of 1 ×  ea
Supelco
SPME Fiber Assembly, matrix active group 50/30µm DVB/CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler, fiber L 1 cm, pk of 3
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, StableFlex, for use with autosampler
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm(CAR/PDMS, needle size 24 ga, for use with manual holder, StableFlex fiber
Supelco
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm(CAR/PDMS, needle size 23 ga, for use with manual holder