- New role of ID3 in melanoma adaptive drug-resistance.
New role of ID3 in melanoma adaptive drug-resistance.
Oncotarget (2018-01-05)
Sachindra, Lionel Larribère, Daniel Novak, Huizi Wu, Laura Hüser, Karol Granados, Elias Orouji, Jochen Utikal
PMID29299138
ABSTRACT
Adaptive resistance to targeted therapy such as BRAF inhibitors represents in melanoma a major drawback to this otherwise powerful treatment. Some of the underlying molecular mechanisms have recently been described: hyperactivation of the BRAF-MAPK pathway, of the AKT pathway, of the TGFβ/EGFR/PDGFRB pathway, or the low MITF/AXL ratio. Nevertheless, the phenomenon of early resistance is still not clearly understood. In this report, we show that knockdown of neural crest-associated gene
MATERIALS