Merck
  • The V3 Loop of HIV-1 Env Determines Viral Susceptibility to IFITM3 Impairment of Viral Infectivity.
The V3 Loop of HIV-1 Env Determines Viral Susceptibility to IFITM3 Impairment of Viral Infectivity.
Journal of virology (2017-01-20)
Yimeng Wang, Qinghua Pan, Shilei Ding, Zhen Wang, Jingyou Yu, Andrés Finzi, Shan-Lu Liu, Chen Liang
ABSTRACT
Interferon-inducible transmembrane proteins (IFITMs) inhibit a broad spectrum of viruses, including HIV-1. IFITM proteins deter HIV-1 entry when expressed in target cells and also impair HIV-1 infectivity when expressed in virus producer cells. However, little is known about how viruses resist IFITM inhibition. In this study, we have investigated the susceptibilities of different primary isolates of HIV-1 to the inhibition of viral infectivity by IFITMs. Our results demonstrate that the infectivity of different HIV-1 primary isolates, including transmitted founder viruses, is diminished by IFITM3 to various levels, with strain AD8-1 exhibiting strong resistance. Further mutagenesis studies revealed that HIV-1 Env, and the V3 loop sequence in particular, determines the extent of inhibition of viral infectivity by IFITM3. IFITM3-sensitive Env proteins are also more susceptible to neutralization by soluble CD4 or the 17b antibody than are IFITM3-resistant Env proteins. Together, data from our study suggest that the propensity of HIV-1 Env to sample CD4-bound-like conformations modulates viral sensitivity to IFITM3 inhibition.
Sigma-Aldrich
Puromycin dihydrochloride from Streptomyces alboniger, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
G 418 disulfate salt, powder, BioReagent, suitable for cell culture
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Anti-HIV-1 P24 antibody produced in rabbit, affinity isolated antibody