Merck
  • Transcriptomic Profiling of Sequential Tumors from Breast Cancer Patients Provides a Global View of Metastatic Expression Changes Following Endocrine Therapy.
Transcriptomic Profiling of Sequential Tumors from Breast Cancer Patients Provides a Global View of Metastatic Expression Changes Following Endocrine Therapy.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-08-05)
Jean McBryan, Ailís Fagan, Damian McCartan, Fiona T Bane, Damir Varešlija, Sinéad Cocchiglia, Christopher Byrne, Jarlath Bolger, Marie McIlroy, Lance Hudson, Paul Tibbitts, Peadar Ó Gaora, Arnold D Hill, Leonie S Young
ABSTRACT
Disease recurrence is a common problem in breast cancer and yet the mechanisms enabling tumor cells to evade therapy and colonize distant organs remain unclear. We sought to characterize global expression changes occurring with metastatic disease progression in the endocrine-resistant setting. Here, for the first time, RNAsequencing has been performed on matched primary, nodal, and liver metastatic tumors from tamoxifen-treated patients following disease progression. Expression of genes commonly elevated in the metastases of sequenced patients was subsequently examined in an extended matched patient cohort with metastatic disease from multiple sites. The impact of tamoxifen treatment on endocrine-resistant tumors in vivo was investigated in a xenograft model. The extent of patient heterogeneity at the gene level was striking. Less than 3% of the genes differentially expressed between sequential tumors were common to all patients. Larger divergence was observed between primary and liver tumors than between primary and nodal tumors, reflecting both the latency to disease progression and the genetic impact of intervening therapy. Furthermore, an endocrine-resistant in vivo mouse model demonstrated that tamoxifen treatment has the potential to drive disease progression and establish distant metastatic disease. Common functional pathways altered during metastatic, endocrine-resistant progression included extracellular matrix receptor interactions and focal adhesions. This novel global analysis highlights the influence of primary tumor biology in determining the transcriptomic profile of metastatic tumors, as well as the need for adaptations in cell-cell communications to facilitate successful tumor cell colonization of distant host organs.
Sigma-Aldrich
Anti-Lamin B2 Antibody, clone LN43, clone LN43, Chemicon®, from mouse
Sigma-Aldrich
Anti-phospho-erbB-2/HER-2 (Tyr1248) Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Active-β-Catenin (Anti-ABC) Antibody, clone 8E7, clone 8E7, Upstate®, from mouse
Sigma-Aldrich
Anti-KRT19 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-MCM4 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution