Merck
  • Alternative Processing of the Amyloid Precursor Protein Family by Rhomboid Protease RHBDL4.
Alternative Processing of the Amyloid Precursor Protein Family by Rhomboid Protease RHBDL4.
The Journal of biological chemistry (2016-08-27)
Sandra Paschkowsky, Mehdi Hamzé, Felix Oestereich, Lisa Marie Munter
ABSTRACT
The amyloid precursor protein (APP) is an ubiquitously expressed cell surface protein and a key molecule in the etiology of Alzheimer disease. Amyloidogenic processing of APP through secretases leads to the generation of toxic amyloid β (Aβ) peptides, which are regarded as the molecular cause of the disease. We report here an alternative processing pathway of APP through the mammalian intramembrane rhomboid protease RHBDL4. RHBDL4 efficiently cleaves APP inside the cell, thus bypassing APP from amyloidogenic processing, leading to reduced Aβ levels. RHBDL4 cleaves APP multiple times in the ectodomain, resulting in several N- and C-terminal fragments that are not further degraded by classical APP secretases. Knockdown of endogenous RHBDL4 results in decreased levels of C-terminal fragments derived from endogenous APP. Similarly, we found the APP family members APLP1 and APLP2 to be substrates of RHBDL4. We conclude that RHBDL4-mediated APP processing provides insight into APP and rhomboid physiology and qualifies for further investigations to elaborate its impact on Alzheimer disease pathology.
Sigma-Aldrich
Anti-APP A4 Antibody, a.a. 66-81 of APP {NT}, clone 22C11, clone 22C11, Chemicon®, from mouse
Sigma-Aldrich
Anti-RHBDD1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution