Merck
  • Serine, but not glycine, supports one-carbon metabolism and proliferation of cancer cells.
Serine, but not glycine, supports one-carbon metabolism and proliferation of cancer cells.
Cell reports (2014-05-13)
Christiaan F Labuschagne, Niels J F van den Broek, Gillian M Mackay, Karen H Vousden, Oliver D K Maddocks
ABSTRACT
Previous work has shown that some cancer cells are highly dependent on serine/glycine uptake for proliferation. Although serine and glycine can be interconverted and either might be used for nucleotide synthesis and one-carbon metabolism, we show that exogenous glycine cannot replace serine to support cancer cell proliferation. Cancer cells selectively consumed exogenous serine, which was converted to intracellular glycine and one-carbon units for building nucleotides. Restriction of exogenous glycine or depletion of the glycine cleavage system did not impede proliferation. In the absence of serine, uptake of exogenous glycine was unable to support nucleotide synthesis. Indeed, higher concentrations of glycine inhibited proliferation. Under these conditions, glycine was converted to serine, a reaction that would deplete the one-carbon pool. Providing one-carbon units by adding formate rescued nucleotide synthesis and growth of glycine-fed cells. We conclude that nucleotide synthesis and cancer cell proliferation are supported by serine--rather than glycine--consumption.
Sigma-Aldrich
Anti-GLDC antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-SHMT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution