Merck
  • Prooxidant effect of α-tocopherol on soybean oil. Global monitoring of its oxidation process under accelerated storage conditions by
Prooxidant effect of α-tocopherol on soybean oil. Global monitoring of its oxidation process under accelerated storage conditions by
Food chemistry (2017-12-31)
A S Martin-Rubio, P Sopelana, M L Ibargoitia, María D Guillén
ABSTRACT
The effect of adding α-tocopherol in proportions ranging from 0.002 to 5% in weight on the oxidative stability of soybean oil was studied. For the first time, the oxidation process under accelerated storage conditions including evolution of the molar percentages of the several types of oil acyl groups, and formation and evolution of various kinds of oxidation products comprising hydroperoxides, hydroxy-dienes and other alcohols, epoxides, aldehydes and keto-dienes, was followed by
Sigma-Aldrich
trans,trans-2,4-Heptandienal, ≥88%
Sigma-Aldrich
Glyceryl trilinolenate, ≥97% (TLC), liquid
Sigma-Aldrich
trans-2-Penten-1-ol, 95%