Merck
  • Glycan microarray reveal induced IgGs repertoire shift against a dietary carbohydrate in response to rabbit anti-human thymocyte therapy.
Glycan microarray reveal induced IgGs repertoire shift against a dietary carbohydrate in response to rabbit anti-human thymocyte therapy.
Oncotarget (2018-01-20)
Ron Amon, Shani Leviatan Ben-Arye, Limor Engler, Hai Yu, Noha Lim, Ludmilla Le Berre, Kristina M Harris, Mario R Ehlers, Stephen E Gitelman, Xi Chen, Jean-Paul Soulillou, Vered Padler-Karavani
ABSTRACT
Humans have circulating antibodies against diverse glycans containing
Sigma-Aldrich
1,3-Benzodioxole, 99%
Supelco
Discovery® Glycan SPE Tube, bed wt. 1 g, volume 12 mL, pk of 20
Supelco
Discovery® Glycan SPE Tube, 5 g, volume 60 mL, pk of 16
Supelco
Discovery® Glycan SPE Bulk Packing, pkg of 50 g
Supelco
Discovery® Glycan SPE Tube, bed wt. 500 mg, volume 6 mL, pk of 30
Supelco
Discovery® Glycan SPE Tube, bed wt. 250 mg, volume 3 mL, pk of 54