Merck
  • Carbon Catabolite Repression Regulates the Production of the Unique Volatile Sodorifen of
Carbon Catabolite Repression Regulates the Production of the Unique Volatile Sodorifen of
Frontiers in microbiology (2018-01-10)
Nancy Magnus, Teresa Weise, Birgit Piechulla
ABSTRACT
Microorganisms are capable of synthesizing a plethora of secondary metabolites including the long-overlooked volatile organic compounds. Little knowledge has been accumulated regarding the regulation of the biosynthesis of such mVOCs. The emission of the unique compound sodorifen of
Sigma-Aldrich
Nonyl acetate, ≥97%, FCC