Merck
Cell-Surface Displayed Expression of Trehalose Synthase from
Frontiers in microbiology (2018-01-10)
Shaojie Yang, Xin Lv, Xihui Wang, Junqing Wang, Ruiming Wang, Tengfei Wang
ABSTRACT
Yeast cell-surface display technologies have been widely applied in the fields of food, medicine, and feed enzyme production, including lipase, α-amylase, and endoglucanase. In this study, a
Sigma-Aldrich
NSC 632839, ≥98% (HPLC)