Merck
  • Expression of the interferon-inducible chemokines MuMig and Crg-2 following vaccinia virus infection in vivo.
Expression of the interferon-inducible chemokines MuMig and Crg-2 following vaccinia virus infection in vivo.
Immunology and cell biology (2000-04-13)
S Mahalingam, G Karupiah
ABSTRACT
MuMig (monokine induced by gamma interferon) and Crg-2 (cytokine responsive gene) are chemokines of the CXC subfamily. They share activity as T and NK cell chemoattractants. Crg-2 has been shown to be inducible by IFN, TNF, IL-1 and LPS, whereas the expression of MuMig is thought to be strictly dependent on IFN-gamma. In the present study, the kinetics of expression of the genes for MuMig and Crg-2 were analysed by northern blot analysis in organs of normal mice and in groups of gene knockout mice deficient in IFN-gamma (IFN-gamma-/-) or receptors for IFN-alpha/beta and IFN-gamma (double R-/-; DR-/-) after vaccinia virus infection. MuMig mRNA was not expressed in IFN-gamma-/- mice in all organs examined, whereas Crg-2 mRNA levels were marginally reduced. In contrast, MuMig and Crg-2 mRNA transcripts were completely abolished in the DR-/- mice.
Product Description
SAFC
CRG-2 from mouse, BioReagent, recombinant, expressed in E. coli, ≥97% (SDS-PAGE), lyophilized powder, suitable for cell culture