Merck
  • Antibody-mediated blockade of JMJD6 interaction with collagen I exerts antifibrotic and antimetastatic activities.
Antibody-mediated blockade of JMJD6 interaction with collagen I exerts antifibrotic and antimetastatic activities.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2017-08-10)
Silvia Miotti, Alessandro Gulino, Renata Ferri, Mariella Parenza, Agnieszka Chronowska, Daniele Lecis, Sabina Sangaletti, Elda Tagliabue, Claudio Tripodo, Mario P Colombo
ABSTRACT
JMJD6 is known to localize in the nucleus, exerting histone arginine demethylase and lysyl hydroxylase activities. A novel localization of JMJD6 in the extracellular matrix, resulting from its secretion as a soluble protein, was unveiled by a new anti-JMJD6 mAb called P4E11, which was developed to identify new targets in the stroma. Recombinant JMJD6 binds with collagen type I (Coll-I), and distinct JMJD6 peptides interfere with collagen fibrillogenesis, collagen-fibronectin interaction, and adhesion of human tumor cells to the collagen substrate. P4E11 and collagen binding to JMJD6 are mutually exclusive because the amino acid sequences of JMJD6 necessary for the interaction with Coll-I are part of the conformational epitope recognized by P4E11. In mice injected with mouse 4T1 breast carcinoma cells, treatment with P4E11 reduced fibrosis at the primary tumor and prevented lung metastases. Reduction of fibrosis has also been documented in human breast and ovarian tumors (MDA-MB-231 and IGROV1, respectively) xenotransplanted into immunodeficient mice treated with P4E11. In summary, this study uncovers a new localization and function for JMJD6 that is most likely independent from its canonical enzymatic activities, and demonstrates that JMJD6 can functionally interact with Coll-I. P4E11 mAb, inhibiting JMJD6/Coll-I interaction, represents a new opportunity to target fibrotic and tumor diseases.-Miotti, S., Gulino, A., Ferri, R., Parenza, M., Chronowska, A., Lecis, D., Sangaletti, S., Tagliabue, E., Tripodo, C., Colombo, M. P. Antibody-mediated blockade of JMJD6 interaction with collagen I exerts antifibrotic and antimetastatic activities.
Sigma-Aldrich
Anti-Phosphatidylserine Receptor antibody produced in rabbit, 1-1.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-Collagen, Type I antibody produced in mouse, clone COL-1, ascites fluid
Sigma-Aldrich
JMJD6 human, recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)
Sigma-Aldrich
Anti-Mouse Collagen Type I Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Fibronectin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution