Merck
  • Antioxidant capacity and major polyphenol composition of teas as affected by geographical location, plantation elevation and leaf grade.
Antioxidant capacity and major polyphenol composition of teas as affected by geographical location, plantation elevation and leaf grade.
Food chemistry (2017-11-10)
Chuang Zhang, Claire Li-Chieh Suen, Chao Yang, Siew Young Quek
ABSTRACT
Tea polyphenols have been a topic of discussion due to their health benefits. Nevertheless, detailed studies on the antioxidant capacity and polyphenol contents of teas in relation to factors including geographical locations, plantation elevations and leaf grades have been limited. In this study, 53 tea samples were analysed to determine the individual and total catechin and theaflavin contents by HPLC and the total antioxidant capacity by Oxygen Radical Absorbance Capacity (ORAC) methods. Results show that the polyphenol (catechins and theaflavins) contents were significantly influenced by plantation location. Black tea from low plantation elevation contained 22-28% more polyphenols than those from high elevation. Small tea leaves had up to 15% more polyphenols than larger leaves from similar elevation. The results were further confirmed by Principal Composition Analysis (PCA), which grouped the black and green tea samples into 3 different clusters, respectively.
Sigma-Aldrich
(−)-Catechin, ≥97% (HPLC), from green tea