Merck
H3.3
Cancer cell (2017-11-07)
Manav Pathania, Nicolas De Jay, Nicola Maestro, Ashot S Harutyunyan, Justyna Nitarska, Pirasteh Pahlavan, Stephen Henderson, Leonie G Mikael, Angela Richard-Londt, Ying Zhang, Joana R Costa, Steven Hébert, Sima Khazaei, Nisreen Samir Ibrahim, Javier Herrero, Antonella Riccio, Steffen Albrecht, Robin Ketteler, Sebastian Brandner, Claudia L Kleinman, Nada Jabado, Paolo Salomoni
ABSTRACT
Gain-of-function mutations in histone 3 (H3) variants are found in a substantial proportion of pediatric high-grade gliomas (pHGG), often in association with TP53 loss and platelet-derived growth factor receptor alpha (PDGFRA) amplification. Here, we describe a somatic mouse model wherein H3.3
Sigma-Aldrich
MISSION® esiRNA, targeting human ATRX
Buprenorphine, European Pharmacopoeia (EP) Reference Standard