Merck
  • Hypothermic machine perfusion with metformin-University of Wisconsin solution for
Hypothermic machine perfusion with metformin-University of Wisconsin solution for
World journal of gastroenterology (2017-11-17)
Yi-Chao Chai, Guo-Xin Dang, Hai-Qi He, Jian-Hua Shi, Hong-Ke Zhang, Rui-Tao Zhang, Bo Wang, Liang-Shuo Hu, Yi Lv
ABSTRACT
To compare the effect of University of Wisconsin (UW) solution with or without metformin, an AMP-activated protein kinase (AMPK) activator, for preserving standard and marginal liver grafts of young and aged rats Eighteen young (4 mo old) and 18 aged (17 mo old) healthy male SD rats were selected and randomly divided into three groups: control group, UW solution perfusion group (UWP), and UW solution with metformin perfusion group (MUWP). Aspartate aminotransferase (AST), alanine aminotransferase (ALT), lactate dehydrogenase (LDH), interleukin-18 (IL-18), and tumor necrosis factor-alpha (TNF-α) in the perfused liquid were tested. The expression levels of AMPK and endothelial nitric oxide synthase (eNOS) in liver sinusoidal endothelial cells were also examined. Additionally, microscopic evaluation of the harvested perfused liver tissue samples was done. AST, ALT, LDH, IL-18 and TNF-α levels in the young and aged liver-perfused liquid were, respectively, significantly lower in the MUWP group than in the UWP group ( The addition of metformin to the UW preservative solution for