Merck
Cell cycle-dependent regulation of cytoglobin by Skp2.
FEBS letters (2017-09-28)
Rince John, Yama Atri, Vaibhav Chand, Neha Jaiswal, Kritika Raj, Alo Nag
ABSTRACT
Cytoglobin (Cygb) is a cellular haemoprotein belonging to the globin family with ambiguous biological functions. Downregulation of Cygb in many cancers is indicative of its tumour-suppressive role. This is the first report showing the cell cycle regulation of Cygb, which was found to peak at G1 and rapidly decline in S phase. Importantly, Skp2-mediated degradation of Cygb was identified as the key mechanism for controlling its oscillating levels during the cell cycle. Moreover, overexpression of Cygb stimulates hypophosphorylation of Rb causing delayed cell cycle progression. Overall, the study reveals a novel mechanism for the regulated expression of Cygb and also assigns a new role to Cygb in cell cycle control.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Kininogen high molecular weight, Two Chain from human plasma, lyophilized powder