Merck
  • Basic Fibroblast Growth Factor 2 Is a Determinant of CD4 T Cell-Airway Smooth Muscle Cell Communication through Membrane Conduits.
Basic Fibroblast Growth Factor 2 Is a Determinant of CD4 T Cell-Airway Smooth Muscle Cell Communication through Membrane Conduits.
Journal of immunology (Baltimore, Md. : 1950) (2017-09-20)
Soroor Farahnak, Toby K McGovern, Rachael Kim, Michael O'Sullivan, Brian Chen, Minhyoung Lee, Haruka Yoshie, Anna Wang, Joyce Jang, Saba Al Heialy, Anne-Marie Lauzon, James G Martin
ABSTRACT
Activated CD4 T cells connect to airway smooth muscle cells (ASMCs) in vitro via lymphocyte-derived membrane conduits (LMCs) structurally similar to membrane nanotubes with unknown intercellular signals triggering their formation. We examined the structure and function of CD4 T cell-derived LMCs, and we established a role for ASMC-derived basic fibroblast growth factor 2 (FGF2b) and FGF receptor (FGFR)1 in LMC formation. Blocking FGF2b's synthesis and FGFR1 function reduced LMC formation. Mitochondrial flux from ASMCs to T cells was partially FGF2b and FGFR1 dependent. LMC formation by CD4 T cells and mitochondrial transfer from ASMCs was increased in the presence of asthmatic ASMCs that expressed more mRNA for FGF2b compared with normal ASMCs. These observations identify ASMC-derived FGF2b as a factor needed for LMC formation by CD4 T cells, affecting intercellular communication.
Sigma-Aldrich
PD-161570, ≥98% (HPLC)