Merck
  • Benefits of Genomic Insights and CRISPR-Cas Signatures to Monitor Potential Pathogens across Drinking Water Production and Distribution Systems.
Benefits of Genomic Insights and CRISPR-Cas Signatures to Monitor Potential Pathogens across Drinking Water Production and Distribution Systems.
Frontiers in microbiology (2017-11-04)
Ya Zhang, Masaaki Kitajima, Andrew J Whittle, Wen-Tso Liu
ABSTRACT
The occurrence of pathogenic bacteria in drinking water distribution systems (DWDSs) is a major health concern, and our current understanding is mostly related to pathogenic species such as
Sigma-Aldrich
Cyclopropane, ≥99%