Merck
The
Frontiers in plant science (2017-11-23)
Yi Zhang, Yuhan Gao, Yingbo Liang, Yijie Dong, Xiufen Yang, Jingjing Yuan, Dewen Qiu
ABSTRACT
During pathogenic infection, hundreds of proteins that play vital roles in the
Sigma-Aldrich
Chitinase from Streptomyces griseus, lyophilized powder (essentially salt free), ≥200 units/g solid
Sigma-Aldrich
Chitin from shrimp shells, suitable for analysis of chitinase, BioReagent, purified powder