Merck
  • Study of ChiR function in Serratia marcescens and its application for improving 2,3-butanediol from crystal chitin.
Study of ChiR function in Serratia marcescens and its application for improving 2,3-butanediol from crystal chitin.
Applied microbiology and biotechnology (2017-09-09)
Qiang Yan, Eunsoo Hong, Stephen S Fong
ABSTRACT
Microbial utilization of chitin, a potential renewable biomass feedstock, is being pursued as a means of developing novel consolidated bioprocessing for the production of chemicals. Serratia marcescens is a gram-negative bacterium that is known for its chitinolytic capability and as a native 2,3-butanediol producer. In S. marcescens, ChiR has been suggested to be a positive regulator of chitinase production. In this study, we aim to understand the effect of ChiR in regulating nine chitinase-related genes in S. marcescens Db11 and demonstrate manipulation of chiR as a useful and efficient genetic target to enhance chitin utilization. First, a chiR overexpression (chiROE) strain and a chiR deletion (ΔchiR) strain were generated and characterized in terms of cellular growth, chitinase activity, and total secreted protein. Compared to the wild-type Db11 strain, the S. marcescens chiROE strain showed an increase in chitinase activity (2.14- to 6.31-fold increase). Increased transcriptional expression of chitinase-related genes was measured using real-time PCR, showing 2.12- to 10.93-fold increases. The S. marcescens ΔchiR strain showed decreases in chitinase activity (4.5- to 25-fold decrease), confirming ChiR's role as a positive regulator of chitinase expression. Finally, chiR overexpression was investigated as a means of increasing biochemical production (2,3-butanediol) from crystal chitin. The chiROE strain produced 1.13 ± 0.08 g/L 2,3-butanediol from 2% crystal chitin, a 2.83-fold improvement from the wild-type strain, indicating ChiR is an important and useful genetic engineering target for enhancing chitin utilization in S. marcescens.
Sigma-Aldrich
Chitinase Assay Kit, sufficient for 100 multiwell tests
Pricing and availability is not currently available.
Sigma-Aldrich
Chitin from shrimp shells, practical grade, powder
Pricing and availability is not currently available.