Merck
In-frame seven amino-acid duplication in
European journal of endocrinology (2017-06-22)
Roberto Salvatori, Serban Radian, Yoan Diekmann, Donato Iacovazzo, Alessia David, Plamena Gabrovska, Giorgia Grassi, Anna-Marie Bussell, Karen Stals, Astrid Weber, Richard Quinton, Elizabeth C Crowne, Valentina Corazzini, Lou Metherell, Tara Kearney, Daniel Du Plessis, Ajay Kumar Sinha, Atik Baborie, Anne-Lise Lecoq, Philippe Chanson, Olaf Ansorge, Sian Ellard, Peter J Trainer, David Balding, Mark G Thomas, Márta Korbonits
ABSTRACT
Mutations in the aryl hydrocarbon receptor-interacting protein ( Observational, inferential and experimental study, including: Nine European-origin, unrelated c.805_825dup-positive pedigrees (four familial, five sporadic from the UK, USA and France) included 16 affected (nine gigantism/four acromegaly/two non-functioning pituitary adenoma patients and one prospectively diagnosed acromegaly patient) and nine unaffected carriers. All pedigrees shared a 2.79 Mbp haploblock around The c.805_825dup allele, originating from a common ancestor, associates with a severe clinical phenotype and a high frequency of gigantism. The mutation is likely to be the result of slipped strand mispairing and affects protein-protein interactions and AIP protein stability.
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Anti-HA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
IgG from mouse serum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder