Merck
  • Determination of acetamiprid, imidacloprid, and spirotetramat and their relevant metabolites in pistachio using modified QuEChERS combined with liquid chromatography-tandem mass spectrometry.
Determination of acetamiprid, imidacloprid, and spirotetramat and their relevant metabolites in pistachio using modified QuEChERS combined with liquid chromatography-tandem mass spectrometry.
Food chemistry (2017-09-28)
Mohammad Faraji, Roya Noorbakhsh, Hooshang Shafieyan, Mohammadkazem Ramezani
ABSTRACT
A QuEChERS based methodology was developed for the simultaneous identification and quantification of acetamiprid, imidacloprid, and spirotetramat and their relevant metabolites in pistachio by liquid chromatography-tandem mass spectrometry for the first time. First, sample extraction was done with MeCN:citrate buffer:NaHCO
Product Description
Supelco
Imidacloprid-d4, PESTANAL®, analytical standard
Supelco
Spirotetramat, PESTANAL®, analytical standard
Supelco
Supel QuE, Citrate/Sodium Bicarbonate Tube, pk of 50
Supelco
Acetamiprid-N-desmethyl, PESTANAL®, analytical standard