Merck
  • Oligodendroglial excitability mediated by glutamatergic inputs and Nav1.2 activation.
Oligodendroglial excitability mediated by glutamatergic inputs and Nav1.2 activation.
Nature communications (2017-09-17)
Emmanuelle Berret, Tara Barron, Jie Xu, Emily Debner, Eun Jung Kim, Jun Hee Kim
ABSTRACT
Oligodendrocyte (OL) maturation and axon-glial communication are required for proper myelination in the developing brain. However, physiological properties of OLs remain largely uncharacterized in different brain regions. The roles of oligodendroglial voltage-activated Na+ channels (Nav) and electrical excitability in relation to maturation to the myelinating stage are controversial, although oligodendroglial excitability is potentially important for promoting axon myelination. Here we show spiking properties of OLs and their role in axon-glial communication in the auditory brainstem. A subpopulation of pre-myelinating OLs (pre-OLs) can generate Nav1.2-driven action potentials throughout postnatal development to early adulthood. In addition, excitable pre-OLs receive glutamatergic inputs from neighboring neurons that trigger pre-OL spikes. Knockdown of Nav1.2 channels in pre-OLs alters their morphology, reduces axon-OL interactions and impairs myelination. Our results suggest that Nav1.2-driven spiking of pre-OLs is an integral component of axon-glial communication and is required for the function and maturation of OLs to promote myelination.Axon-glial communication is important for myelination. Here the authors show that during postnatal development in rats, a subpopulation of pre-myelinating oligodendrocytes in the auditory brainstem receive excitatory inputs and can generate Nav 1.2-driven action potentials, and that such process promotes myelination.
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Olig1 Antibody, ascites fluid, Chemicon®
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 1 Antibody, serum, Chemicon®