Merck
  • KCa3.1 Inhibition Switches the Astrocyte Phenotype during Astrogliosis Associated with Ischemic Stroke Via Endoplasmic Reticulum Stress and MAPK Signaling Pathways.
KCa3.1 Inhibition Switches the Astrocyte Phenotype during Astrogliosis Associated with Ischemic Stroke Via Endoplasmic Reticulum Stress and MAPK Signaling Pathways.
Frontiers in cellular neuroscience (2017-10-28)
Zhihua Yu, Mengni Yi, Tianjiao Wei, Xiaoling Gao, Hongzhuan Chen
ABSTRACT
Ischemic stroke is a devastating neurological disease that can initiate a phenotype switch in astrocytes. Reactive astrogliosis is a significant pathological feature of ischemic stroke and is accompanied by changes in gene expression, hypertrophied processes and proliferation. The intermediate-conductance Ca
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid