Merck
  • Gas chromatography-mass spectrometry and high-performance liquid chromatography-diode array detection for dating of paper ink.
Gas chromatography-mass spectrometry and high-performance liquid chromatography-diode array detection for dating of paper ink.
Journal of chromatography. A (2017-08-16)
Oscar Díaz-Santana, Daura Vega-Moreno, Francisco Conde-Hardisson
ABSTRACT
An extraction and determination method is shown for the analysis of dyes and solvents present in two types of ballpoint pen inks that are deposited onto paper. Ink extracts are analysed using a combination of gas chromatography with mass spectrometry (GC-MS), and high-pressure liquid chromatography with photodiode array detection (HPLC-DAD), within a single sample extraction procedure. Seventeen solvents and thirteen dyes contained in two Montblanc
Sigma-Aldrich
4-Ethylaniline, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)