Merck
  • Determination of rat oral bioavailability of soy-derived phytoestrogens using an automated on-column extraction procedure and electrospray tandem mass spectrometry.
Determination of rat oral bioavailability of soy-derived phytoestrogens using an automated on-column extraction procedure and electrospray tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2003-10-14)
Larry M Mallis, Ani B Sarkahian, Heather A Harris, Mei-Yi Zhang, Oliver J McConnell
ABSTRACT
In recent years, consumption of herbal supplements as an alternative to pharmaceutical drug therapy has increased. For example, with the health claims labeling which describes the link between soy-protein and a reduced risk of coronary heart disease (CHD), the consumption of soy and soy-derived phytoestrogens has increased dramatically. That being said, the oral bioavailability of only a few soy phytoestrogens such as Daidzein and Genestein have been previously estimated. In this paper, we present the calculated percent of rat oral bioavailability of five soy-derived phytoestrogens (Genistein, Daidzein, Biochanin A, Coumestrol, and Zearalenone) in male Sprague-Dawley rats. The plasma quantitation required for the bioavailability calculation is performed by using a rapid on-line plasma extraction procedure for the quantitative analysis. To further speed up the analysis the rats were dosed using the 'n-in-one' (cassette) protocol. The rapid on-line extraction/quantitation methodology coupled to the cassette dosing analysis of phytoestrogens is the key point of this paper. The limit of quantitation (LOQ) for each compound was 1-1000 ng/ml with each plasma sample analysis taking less than 2 min. In general the percent oral bioavailability was determined to be between 11 and 28%.
Sigma-Aldrich
Coumestrol, ≥95.0% (HPLC)
Pricing and availability is not currently available.