Merck
  • The isozyme selective phosphodiesterase-4 inhibitor, ABI-4, attenuates the effects of lipopolysaccharide in human cells and rodent models of peripheral and CNS inflammation.
The isozyme selective phosphodiesterase-4 inhibitor, ABI-4, attenuates the effects of lipopolysaccharide in human cells and rodent models of peripheral and CNS inflammation.
Brain, behavior, and immunity (2017-04-26)
Joseph R Hedde, Ashley N Hanks, Christopher J Schmidt, Zoë A Hughes
ABSTRACT
Inhibitors of phosphodiesterase-4 (PDE4) have been approved for the treatment of inflammatory disorders, but are associated with dose-limiting nausea and vomiting. These side effects are hypothesized to be mediated by inhibition of the PDE4D isozyme. Here we demonstrate the anti-inflammatory effects of the novel brain penetrant PDE4D-sparing PDE4 inhibitor, ABI-4. ABI-4 was a potent (EC
Sigma-Aldrich
4′-Chlorodiazepam, ≥98% (TLC)
Sigma-Aldrich
Rolipram, solid, ≥98% (HPLC)