Merck
  • Role of Surface Capping Molecule Polarity on the Optical Properties of Solution Synthesized Germanium Nanocrystals.
Role of Surface Capping Molecule Polarity on the Optical Properties of Solution Synthesized Germanium Nanocrystals.
Langmuir : the ACS journal of surfaces and colloids (2017-05-30)
B F P McVey, P B O'Mara, A J McGrath, A Faramus, V B Yasarapudi, V R Gonçales, V T G Tan, T W Schmidt, J J Gooding, R D Tilley
ABSTRACT
The role surface capping molecules play in dictating the optical properties of semiconductor nanocrystals (NCs) is becoming increasingly evident. In this paper the role of surface capping molecule polarity on the optical properties of germanium NCs (Ge NCs) is explored. Capping molecules are split into two groups: nonpolar and polar. The NCs are fully characterized structurally and optically to establish the link between observed optical properties and surface capping molecules. Ge NC optical properties altered by surface capping molecule polarity include emission maximum, emission lifetime, quantum yield, and Stokes shift. For Ge NCs, this work also allows rational tuning of their optical properties through changes to surface capping molecule polarity, leading to improvements in emerging Ge based bioimaging and optoelectronic devices.
Sigma-Aldrich
1,7-Octadiene, 98%