Merck
  • The Candida albicans transcription factor Cas5 couples stress responses, drug resistance and cell cycle regulation.
The Candida albicans transcription factor Cas5 couples stress responses, drug resistance and cell cycle regulation.
Nature communications (2017-09-13)
Jinglin L Xie, Longguang Qin, Zhengqiang Miao, Ben T Grys, Jacinto De La Cruz Diaz, Kenneth Ting, Jonathan R Krieger, Jiefei Tong, Kaeling Tan, Michelle D Leach, Troy Ketela, Michael F Moran, Damian J Krysan, Charles Boone, Brenda J Andrews, Anna Selmecki, Koon Ho Wong, Nicole Robbins, Leah E Cowen
ABSTRACT
The capacity to coordinate environmental sensing with initiation of cellular responses underpins microbial survival and is crucial for virulence and stress responses in microbial pathogens. Here we define circuitry that enables the fungal pathogen Candida albicans to couple cell cycle dynamics with responses to cell wall stress induced by echinocandins, a front-line class of antifungal drugs. We discover that the C. albicans transcription factor Cas5 is crucial for proper cell cycle dynamics and responses to echinocandins, which inhibit β-1,3-glucan synthesis. Cas5 has distinct transcriptional targets under basal and stress conditions, is activated by the phosphatase Glc7, and can regulate the expression of target genes in concert with the transcriptional regulators Swi4 and Swi6. Thus, we illuminate a mechanism of transcriptional control that couples cell wall integrity with cell cycle regulation, and uncover circuitry governing antifungal drug resistance.Cas5 is a transcriptional regulator of responses to cell wall stress in the fungal pathogen Candida albicans. Here, Xie et al. show that Cas5 also modulates cell cycle dynamics and responses to antifungal drugs.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)