Merck
  • Formation of multilayer ultrathin assemblies using chemical lithography.
Formation of multilayer ultrathin assemblies using chemical lithography.
Langmuir : the ACS journal of surfaces and colloids (2010-04-21)
Chuanzhen Zhou, Amy V Walker
ABSTRACT
Ultrathin complex multilayer structures have many potential applications in molecular and organic electronics, sensing, biotechnology and other areas. Reported here is a method by which to construct multifunctional, multilayer, patterned structures, using alkanethiolate SAMs adsorbed on Au, UV photopatterning, and chemoselective covalent bond formation. We demonstrate that amide coupling is efficient for producing multilayer structures on -COOH-terminated SAMs, while oxime coupling is efficient for producing multilayer structures on -CHO-terminated SAMs. Reaction yields obtained are approximately 67% and approximately 84% for the coupling of the first layer (bilayer formation) for amide and oxime coupling, respectively. Subsequent adlayer formation occurs with approximately 100% yield in both cases. The resulting adlayers are chemically robust and are suitable for subsequent chemical processing. Finally, both chemistries are used to produce a complex multilayer structure atop a UV photopatterned SAM. The resulting construct is well-defined and has the same lateral resolution as the photopatterned SAM substrate. The method demonstrated here is synthetically flexible and allows for the assembly of functionally complex surfaces and, in principle, the incorporation of biomolecules and metals.
Sigma-Aldrich
16-Mercaptohexadecanoic acid, 98%