Merck
  • Optimal determination of steric mass action model parameters for beta-lactoglobulin using static batch experiments.
Optimal determination of steric mass action model parameters for beta-lactoglobulin using static batch experiments.
Journal of chromatography. A (2010-05-07)
Tilman Barz, Verena Löffler, Harvey Arellano-Garcia, Günter Wozny
ABSTRACT
In this work, parameters of the steric mass-formalism SMA are optimally ascertained for a reliable determination of the adsorption isotherms of beta-lactoglobulin A and B under non-isocratic conditions. For this purpose, static batch experiments are used in contrast to the protocols based on different experimental steps, which use a chromatographic column. It is shown that parameters can already be determined for a small number of experiments by using a systematic procedure based on optimal model-based experimental design and an efficient NLP-solver. The in different works observed anti-Langmuir shape of the isotherm for small concentrations of beta-lactoglobulin A was corroborated. Moreover, we also found indications for a porosity variation with changing protein concentrations.
Sigma-Aldrich
β-Lactoglobulin B from bovine milk, ≥90% (PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
β-Lactoglobulin from bovine milk, ≥90% (PAGE), lyophilized powder
Pricing and availability is not currently available.