Merck
  • Uncovering the effect of low-frequency static magnetic field on tendon-derived cells: from mechanosensing to tenogenesis.
Uncovering the effect of low-frequency static magnetic field on tendon-derived cells: from mechanosensing to tenogenesis.
Scientific reports (2017-09-10)
Tamagno Pesqueira, Raquel Costa-Almeida, Manuela E Gomes
ABSTRACT
Magnetotherapy has been receiving increased attention as an attractive strategy for modulating cell physiology directly at the site of injury, thereby providing the medical community with a safe and non-invasive therapy. Yet, how magnetic field influences tendon cells both at the cellular and molecular levels remains unclear. Thus, the influence of a low-frequency static magnetic field (2 Hz, 350 mT) on human tendon-derived cells was studied using different exposure times (4 and 8 h; short-term studies) and different regimens of exposure to an 8h-period of magnetic stimulation (continuous, every 24 h or every 48 h; long-term studies). Herein, 8 h stimulation in short-term studies significantly upregulated the expression of tendon-associated genes SCX, COL1A1, TNC and DCN (p < 0.05) and altered intracellular Ca2+ levels (p < 0.05). Additionally, every 24 h regimen of stimulation significantly upregulated COL1A1, COL3A1 and TNC at day 14 in comparison to control (p < 0.05), whereas continuous exposure differentially regulated the release of the immunomodulatory cytokines IL-1β and IL-10 (p < 0.001) but only at day 7 in comparison to controls. Altogether, these results provide new insights on how low-frequency static magnetic field fine-tune the behaviour of tendon cells according to the magnetic settings used, which we foresee to represent an interesting candidate to guide tendon regeneration.
Sigma-Aldrich
Fluo-3, suitable for fluorescence, ~70%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone hVIN-1, ascites fluid
Sigma-Aldrich
Phalloidin–Tetramethylrhodamine B isothiocyanate, sequence from Amanita phalloides(synthetic: peptide sequence)
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Sigma-Aldrich
Fluo 3-AM solution, 1 mM in DMSO, ≥99.0% (TLC)