Merck
  • Free immunoglobulin light chain (FLC) promotes murine colitis and colitis-associated colon carcinogenesis by activating the inflammasome.
Free immunoglobulin light chain (FLC) promotes murine colitis and colitis-associated colon carcinogenesis by activating the inflammasome.
Scientific reports (2017-07-14)
Junfan Ma, Dongyang Jiang, Xiaoting Gong, Wenwei Shao, Zhu Zhu, Weiyan Xu, Xiaoyan Qiu
ABSTRACT
Numerous studies have demonstrated that free Ig light chain (FLC), a novel inflammation mediator, participates in many inflammatory diseases by activating mast cells and extending the survival of neutrophils. However, it remains unclear whether FLC is involved in colitis and colitis-associated colon carcinogenesis (CAC). In this study, we found a significant increase in FLC in murine models of DSS (Dextran Sulfate Sodium Salt)-induced colitis and CAC compared to controls. Peptide F991, a functional blocker of FLC, significantly attenuated colitis progression, which included abrogating the development of diarrhea and tumor burden, elevating survival rate, greatly reducing the infiltration of inflammatory cells (such as ROS
Sigma-Aldrich
Azoxymethane, 13.4 M, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Corn steep solids, spray-dried corn solubles
Pricing and availability is not currently available.
Roche
cOmplete Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Pricing and availability is not currently available.