Merck
  • Analysis of lysophosphatidic acid (LPA) receptor and LPA-induced endometrial prostaglandin-endoperoxide synthase 2 expression in the porcine uterus.
Analysis of lysophosphatidic acid (LPA) receptor and LPA-induced endometrial prostaglandin-endoperoxide synthase 2 expression in the porcine uterus.
Endocrinology (2008-08-16)
Heewon Seo, Mingoo Kim, Yohan Choi, Chang-Kyu Lee, Hakhyun Ka
ABSTRACT
Lysophosphatidic acid (LPA), a simple phospholipid-derived mediator with diverse biological actions, acts through the specific G protein-coupled receptors endothelial differentiation gene (EDG) 2, EDG4, EDG7, and GPR23. Recent studies indicate a critical role for LPA receptor signaling in embryo implantation. To understand how LPA acts in the uterus during pregnancy in pigs, we evaluated: 1) spatial and temporal expression of LPA receptors in the uterine endometrium during the estrous cycle and pregnancy and in early-stage concepti, 2) LPA levels in uterine luminal fluids from d 12 of the estrous cycle and pregnancy, 3) effects of steroid hormones on EDG7 mRNA levels, and 4) effects of LPA on prostaglandin-endoperoxide synthase 2 (PTGS2) mRNA levels in the uterine endometrium using explant cultures. Of the four receptors, EDG7 was dominant, and its expression was regulated by pregnancy stage and status. EDG7 expression was highest on d 12 pregnancy, and localized to the luminal and glandular epithelium, and EDG7 mRNA levels were elevated by estrogen in the endometrium. EDG7 expression was also detected in concepti of d 12 and 15. LPA with various fatty acyl groups was present in the uterine lumen on d 12 of both the estrous cycle and pregnancy. LPA increased PTGS2 mRNA abundance in the uterine endometrium. These results indicate that LPA produced in the uterine endometrium may play a critical role in uterine endometrial function and conceptus development through EDG7-mediated PTGS2 expression during implantation and establishment of pregnancy in pigs.
Sigma-Aldrich
Progesterone, ≥99%
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Mifepristone, ≥98%
Sigma-Aldrich
apo-Transferrin bovine, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
β-Estradiol, ≥98%
Sigma-Aldrich
Oleoyl-L-α-lysophosphatidic acid sodium salt, ≥98%, solid