Merck
  • The endothelial transcription factor ERG mediates Angiopoietin-1-dependent control of Notch signalling and vascular stability.
The endothelial transcription factor ERG mediates Angiopoietin-1-dependent control of Notch signalling and vascular stability.
Nature communications (2017-07-12)
A V Shah, G M Birdsey, C Peghaire, M E Pitulescu, N P Dufton, Y Yang, I Weinberg, L Osuna Almagro, L Payne, J C Mason, H Gerhardt, R H Adams, A M Randi
ABSTRACT
Notch and Angiopoietin-1 (Ang1)/Tie2 pathways are crucial for vascular maturation and stability. Here we identify the transcription factor ERG as a key regulator of endothelial Notch signalling. We show that ERG controls the balance between Notch ligands by driving Delta-like ligand 4 (Dll4) while repressing Jagged1 (Jag1) expression. In vivo, this regulation occurs selectively in the maturing plexus of the mouse developing retina, where Ang1/Tie2 signalling is active. We find that ERG mediates Ang1-dependent regulation of Notch ligands and is required for the stabilizing effects of Ang1 in vivo. We show that Ang1 induces ERG phosphorylation in a phosphoinositide 3-kinase (PI3K)/Akt-dependent manner, resulting in ERG enrichment at Dll4 promoter and multiple enhancers. Finally, we demonstrate that ERG directly interacts with Notch intracellular domain (NICD) and β-catenin and is required for Ang1-dependent β-catenin recruitment at the Dll4 locus. We propose that ERG coordinates Ang1, β-catenin and Notch signalling to promote vascular stability.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Phosphoserine antibody produced in mouse, clone PSR-45, purified from hybridoma cell culture, buffered aqueous solution
Sigma-Aldrich
MISSION® esiRNA, targeting human ERG
Sigma-Aldrich
Anti-Jagged 1 antibody produced in goat, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
IgG, Rabbit, Rabbit IgG Polyclonal Antibody validated for use in ELISA.