Skip to Content
AU
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
ET
ET
American journal of physiology. Regulatory, integrative and comparative physiology (2017-04-26)
Megan M Wenner, Kelly N Sebzda, Andrew V Kuczmarski, Ryan T Pohlig, David G Edwards
PMID
28438762
ABSTRACT
Endothelin-1 (ET-1) contributes to age-related endothelial dysfunction in men via the ET
Product Description
B157
Sigma-Aldrich
BQ-788, ≥95%, solid
Pricing
B150
Sigma-Aldrich
BQ-123, ≥99%, sodium salt, lyophilized powder
Pricing