Merck
Multiwavelength Light-Responsive Au/B-TiO
ACS nano (2017-06-08)
Bumjin Jang, Ayoung Hong, Ha Eun Kang, Carlos Alcantara, Samuel Charreyron, Fajer Mushtaq, Eva Pellicer, Robert Büchel, Jordi Sort, Sung Sik Lee, Bradley J Nelson, Salvador Pané
ABSTRACT
Conventional photocatalytic micromotors are limited to the use of specific wavelengths of light due to their narrow light absorption spectrum, which limits their effectiveness for applications in biomedicine and environmental remediation. We present a multiwavelength light-responsive Janus micromotor consisting of a black TiO