- Chitin degradation and utilization by virulent Aeromonas hydrophila strain ML10-51K.
Chitin degradation and utilization by virulent Aeromonas hydrophila strain ML10-51K.
Virulent Aeromonas hydrophila (vAh) is one of the most important bacterial pathogens that causes persistent outbreaks of motile Aeromonas septicemia in warm-water fishes. The survivability of this pathogen in aquatic environments is of great concern. The aim of this study was to determine the capability of the vAh strain ML10-51K to degrade and utilize chitin. Genome-wide analysis revealed that ML10-51K encodes a suite of proteins for chitin metabolism. Assays in vitro showed that four chitinases, one chitobiase and one chitin-binding protein were secreted extracellularly and participated in chitin degradation. ML10-51K was shown to be able to use not only N-acetylglucosamine and colloidal chitin but also chitin flakes as sole carbon sources for growth. This study indicates that ML10-51K is a highly chitinolytic bacterium and suggests that the capability of effective chitin utilization could enable the bacterium to attain high densities when abundant chitin is available in aquatic niches.